Learn More
ARSJ Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557679
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DSPGMCGYDL YENDNAAWDY DNGIYSTQMY TQRVQQILAS HNPTKPIFLY IAYQAVHSPL QAPGRYFEHY RSIININRRR YAAMLSCLDE AINNV Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 55%.
ARSJ Polyclonal antibody specifically detects ARSJ in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ARSJ | |
Polyclonal | |
Unconjugated | |
ARSJ | |
9330196J05Rik; Arsj; arylsulfatase family member J; arylsulfatase family, member J; arylsulfatase J; ASJ; D830047F08; RGD1307640; UNQ372/PRO708 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79642 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5FYB0 | |
ARSJ | |
Recombinant protein corresponding to Human ARSJ | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only