missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™

Brand:  Fisher Chemical S003250

Code : 04

Additional Details : Weight : 0.25000kg


Quantity 250mL

 View more versions of this product

Product Code. 12973354

  • £71.00 / 250mL


Estimated Shipment
01-03-2018
Add to basket


Specifications

Chemical Identifiers

1g/L
HNO3

Specifications

1g/L
HNO3
250mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Single-element Standard Solution for ICP. Arsenic (As) 1000mg/l in HNO3 2%

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A

Hazard Statement
  • H350-May cause cancer.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .