missing translation for 'onlineSavingsMsg'
Learn More

Arsenic powder, -20 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11321148
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
7440-38-2
As
74.92
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
5359596
CHEBI:27563
arsenic
[As]
This item is not returnable. View return policy
7440-38-2
As
74.92
MFCD00085309
RQNWIZPPADIBDY-UHFFFAOYSA-N
arsenicals, arsenic, inorganic, arsenic-75, arsen, gray, grey, black, metallic, compounds, colloidal
5359596
CHEBI:27563
arsenic
[As]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black to Silver
UN Number UN1558
Quantity 250 g
Percent Purity 99%
Mesh Size −20 Mesh
Odor Odorless
Form Powder
Assay (metals basis)
Chemical Name or Material Arsenic powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'