missing translation for 'onlineSavingsMsg'
Learn More

Arsenic, Oil based standard solution, Specpure™ As 1000μg/g, Thermo Scientific Chemicals

Catalog Number 15446555
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 g
MDL Number MFCD00085309
Grade Specpure™
Physical Form Liquid
Chemical Name or Material Arsenic, Oil based standard solution, As 1000μg/g

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'