missing translation for 'onlineSavingsMsg'
Learn More

Arsenic(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number 15452417
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1303-33-9
As2S3
246.02
MFCD00003435
CUGMJFZCCDSABL-UHFFFAOYSA-N
diarsenic(3+) trisulfanediide
[S--].[S--].[S--].[As+3].[As+3]
This item is not returnable. View return policy
1303-33-9
As2S3
246.02
MFCD00003435
CUGMJFZCCDSABL-UHFFFAOYSA-N
diarsenic(3+) trisulfanediide
[S--].[S--].[S--].[As+3].[As+3]

Arsenic(III) sulfide is used in the treatment of acute promyelocytic leukemia. It is an inorganic photoresist used in the fabrication of photonic crystals and 3-D nanostructures. It is also used as an acousto-optic material, pigment, tanning agent and chalcogenide glass for infrared optics. Further, it is used as an analytical test for presence of dissimilatory arsenic-reducing bacteria (DARB).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 707°C
Melting Point 300°C
Physical Form Powder
Quantity 25 g
UN Number UN1557
Merck Index 14,806
Solubility Information Insoluble in water.
Formula Weight 246.03
Percent Purity 99.9%
Odor Like rotten eggs
Density 3.43 g/mL
Assay (metals basis)
Chemical Name or Material Arsenic (III) sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'