missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic III Speciation Standard (Assurance™ Grade/For ICP-MS), SPEX CertiPrep™
Click to view available options
Quantity:
125 mL
Packaging:
Poly Bottle
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Arsenic +3
- Speciation Standard
- 125 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Specifications
Packaging | Poly Bottle |
CAS Min % | 0.1, 2.0, 97.9 |
Quantity | 125 mL |
CAS Max % | 0.1, 2.0, 97.9 |
Grade | Assurance |
Chemical Name or Material | Arsenic III Speciation Standard |
Safety and Handling
Product Identifier
- 1000 ug/mL Arsenic (III)
- Danger
- Carcinogenicity Category 1A
- H350-May cause cancer.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : hydrochloric acid, Arsenic from Arsenictrioxide
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'