missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Arsenic(III) ethoxide

Brand:  Alfa Aesar™ 031498.18

Code : NEW

Additional Details : CAS Number : 3141-12-6 Weight : 0.05000kg Transport : UN number : 2929 Chem class : 1 Pack group : II

Quantity 50g

 View more versions of this product

Product Code. 11393897

  • £348.85 / 50g


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Specifications

Chemical Identifiers

Arsenic(III) ethoxide
210.07
UN2929
3141-12-6
MFCD00015120
Triethoxyarsine

Specifications

Arsenic(III) ethoxide
Liquid
MFCD00015120
Triethoxyarsine
166°C
Moisture Sensitive
3141-12-6
210.07
UN2929
1.21g/mL
50g
Videos
SDS