missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Arsenic(III) ethoxide

Brand:  Alfa Aesar™ 031498.04

Code : NEW

Additional Details : CAS Number : 3141-12-6 Weight : 0.00200kg Transport : UN number : 2929 Chem class : 1 Pack group : II

Quantity 2g

 View more versions of this product

Product Code. 11313907

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Arsenic(III) ethoxide
210.07
UN2929
3141-12-6
MFCD00015120
Triethoxyarsine

Specifications

Arsenic(III) ethoxide
Liquid
MFCD00015120
Triethoxyarsine
166°C
Moisture Sensitive
3141-12-6
210.07
UN2929
1.21g/mL
2g
Videos
SDS