missing translation for 'onlineSavingsMsg'
Learn More

Arsenic for AAS 1g/L HCl matrix, Fisher chemical™

Catalog Number p-7083647
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
This item is not returnable. View return policy
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 1g/L ±0.002g/L
Packaging Bottle
Quantity 100 mL
Type Flame Single-element Standard Solution
UN Number 1556
Solvent or Matrix HCl
Physical Form Liquid
Chemical Name or Material Arsenic
compliance-icons
Product Identifier
  • Single-element Standard Solution for AAS. Arsenic (As) 1000mg/l in HCl 2% (B 003)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'