missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Arrow Strips

Brand:  Brady™ 251946

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11796645

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

1m x 60mm
PhotoluminescentþGreen
Provide a clear pathway when the power fails
1Each
Arrow Strips
Videos