Learn More
ARMX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552201
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RDQTKKRMAK PKNRAVAGTG ARARAGLRAG FTIDLGSGFS PPTPVRAEAE DRAQDEASAL DTVGAEAVAP AASSAEAQSG AGSQAQEADG AGVGPKAESV VGAAMASAIA PPPGVTEALG AAEAPAMAGA PKVAEAPREA ETSRA Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 55%.
ARMX2 Polyclonal antibody specifically detects ARMX2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ARMX2 | |
Polyclonal | |
Unconjugated | |
ARMCX2 | |
3230401N03Rik; AI043003; ALEX2; ALEX2 protein; ARM protein lost in epithelial cancers on chromosome X 2; armadillo repeat containing, X-linked 2; armadillo repeat protein ALEX2; armadillo repeat-containing X-linked protein 2; ARMCX2; GASP9; Kiaa0512; Protein ALEX2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9823 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L311 | |
ARMCX2 | |
Recombinant protein corresponding to Human ARMX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only