missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, Super Bright 645, eBioscience™

Armenian Hamster Isotype Control

Brand:  Invitrogen 64-4888-80

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15943654

  • £121.00 / 25µg

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The eBio299Arm monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eBio299Arm antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBio299Arm antibody has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405nm) and emits at 645nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product # SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100μL of cell sample+ 100μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405nm; Emission: 645nm; Laser: Violet Laser

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Specifications

Specifications

Armenian Hamster IgG
Control
0.2mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Armenian Hamster
Affinity chromatography
RUO
Isotype Control
Control, Flow Cytometry
eBio299Arm
Super Bright 645
Liquid
IgG
25μg
4°C, store in dark, DO NOT FREEZE
Not Applicable
Videos
SDS
Documents