Thermo Scientific™ Armadillo™ 96-Well PCR Plate PROMO
Optimize robotic applications with these ultra-rigid 96-well PCR plates with U-bottom wells, available in various colors.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: BC-3396-RMP6240
UNSPSC: 41122102
Additional Details:
Additional Details: Weight: 0.90000kg
Product Code. 15563491
Quantity Price
1 £ 109.0 / Pack of 25
Estimated Shipment date
from Supplier 13-04-2017
Add to basket
Description and Specification
Highlights
  • Optically clear skirt - Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design - Improved well surface area improves seal integrity in automated heat sealing
  • ANSI:SBS footprint - With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high throughput applications
  • Thermostable polycarbonate frame design - Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells - Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering - Integrated frame and well design for improved heat sealing
  • Low profile design - Allows for maximum heat transfer and minimal evaporation when utilizing low volume samples (200 µL working volume)
  • Available in both clear and white coloured wells - Ensures the highest level of sensitivity for all PCR/qPCR reactions
  • Barcoding available - All plates can be provided with standard barcoding or barcoded to customer specifications for fully automated applications