Thermo Scientific™ Armadillo™ 384-Well PCR Plates PROMO
Optimize high throughput robotic PCR and qPCR applications with these uItra-rigid 384-well PCR plates with poylcarbonate frames and polypropylene wells.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: AB-3384-RMP6240
X50 Armadillo 384-Well PCR Plate (Clear, White Wells)
UNSPSC: 41122102
Additional Details:
Additional Details: Weight: 1.40000kg
Product Code. 15553481
Quantity Price
1 £ 257.0 / Pack of 50
Estimated Shipment date
from Supplier 06-02-2017
Add to basket
Description and Specification
Highlights
  • Optically clear skirt - Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design - Improved well surface area improves seal integrity in automated heat sealing
  • ANSI:SBS footprint - With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
  • Thermostable polycarbonate frame design - Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells - Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering - Integrated frame and well design for improved heat sealing
  • Low profile design - Allows for maximum heat transfer and minimal evaporation when utilizing low-volume samples (30 µL working volume)
  • Available in both clear and white colored wells - Ensures the highest level of sensitivity for all PCR/qPCR reactions
  • Barcoding available - All plates are available already barcoded and can be provided barcoded to customer specifications for fully-automated applications