missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550331
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total ARHGAP5 protein.
ARHGAP5 Polyclonal antibody specifically detects ARHGAP5 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
ARHGAP5 | |
Polyclonal | |
Unconjugated | |
ARHGAP5 | |
ARHGAP5; GFI2; growth factor independent 2; LRRGT00098; p100 RasGAP-associated p105 protein; p105 RhoGAP; p190B; p190-B; p190BRhoGAP; Rho GTPase activating protein 5; rho GTPase-activating protein 5; Rho GTPase-activating protein 5-like protein; RHOGAP5; rho-type GTPase-activating protein 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
394 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q13017 | |
ARHGAP5 | |
Full length fusion protein of Human ARHGAP5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |