missing translation for 'onlineSavingsMsg'
Learn More

Argos Technologies™ StarFire Safety Burner Accessories

For use with StarFire ST and StarFire XT Safety Bunsen Burners

Brand:  Camping Gaz 39350KIT

Additional Details : CAS Number : 74-98-6 Weight : 2.20500kg Transport : UN number : 2037 Chem class : 2.1


Product Code. 15242329

  • £327.00 / Pack of 24
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Butane Gas Cartridge
Starfire ST and Starfire XT Safety Bunsen Burners
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Butan

Signal Word
  • Danger

Hazard Category
  • Flammable gas.

Hazard Statement
  • H220-Extremely flammable gas.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
  • P381-Eliminate all ignition sources if safe to do so.
  • P403-Store in a well-ventilated place.

Supplemental information
  • MIXTURE LIST-Contains : Hydrocarbures richesen C3-C4 Gaz de pétrole

SDS
Documents

Documents

Certificates
Special Offers

Special Offers