missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1, clone: sl6arg, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9779-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15507686

  • £57.31 / 25µg


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Description

Description

Description: The monoclonal antibody sl6arg recognizes human Arginase-1, a 35kDa enzyme that that helps eliminate nitrogen by converting L-arginine to urea and L-ornithine which further converts to polyamine. These polyamines are important for cell proliferation and removal of toxins that arise from protein degradation. Expression is found in the liver, neutrophils, as well as myeloid derived suppressor cells (MDSC) and neural stem cells. Expression in macrophages has not been determined in humans, while in mice expression in M2 macrophages is well established. Arginase-1 may be expressed in the myeloid population in breast cancer tumors and is typically found in the majority of hepatocellular carcinomas.Applications Reported: This sl6arg antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, immunohistochemical staining, and immunocytochemistry.Applications Tested: This sl6arg antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using either low or high pH antigen retrieval and can be used at less than or equal to 20 μg/mL. This sl6arg antibody has also been tested by immunocytochemistry of fixed and permeabilized human cells and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Arginase
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P05089
AI type I arginase, arginase 1, liver, arginase I, arginase, liver, arginase-1, liver-type arginase, type I arginase
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
sl6arg
Unconjugated
ARG1
Purified
ARG1
IgG1, kappa
25μg
Store at 2-8°C.
Primary
383
Videos

For Research Use Only.