Learn More
Arginase 1 Monoclonal Antibody (sl6arg), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-9779-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Arginase-1 (ARG1) catalyzes the hydrolysis of L-arginine into ornithine and urea, a critical step for the urea cycle. Defects in ARG1 are the cause of argininemia, an autosomal recessive disorder characterized by hyperammonemia. There are several isoforms of mammalian arginase. The type I isoform, is a cytosolic enzyme and expressed predominantly in the liver as a component of the urea cycle. Hepatocellular carcinoma usually shows higher protein expression of ARG1 than normal liver cells.
Arginase 1 Monoclonal antibody specifically detects Arginase 1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Arginase 1 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P05089 | |
ARG1 | |
Affinity chromatography | |
RUO | |
383 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
sl6arg | |
eFluor 660 | |
ARG1 | |
AI; A-I; AI type I arginase; AI256583; Arg1; Arg-1; Arginase; arginase 1; arginase 1 liver; arginase 1, liver; arginase I; arginase, liver; Arginase1; arginase-1; HGNC:663; Liver Arginase; Liver-type arginase; PGIF; similar to arginase, type I; Type 1 Arginase; type I arginase | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.