Learn More
Argatroban, Tocris Bioscience™
Potent thrombin inhibitor
Brand:  Tocris (Bio-Techne) 163710
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13449843
  • £173.46 / 10mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.
This compound is a mixture of 2 diastereomers (approximate ratio 65/35).
Specifications
Chemical Identifiers
C23H36N6O5S
Specifications
Argatroban
>99%
508.63
74863-84-6
C23H36N6O5S