Learn More
Argatroban, Tocris Bioscience™
Potent thrombin inhibitor
POA
Specifications
Chemical Name or Material | Argatroban |
---|---|
CAS | 74863-84-6 |
Assay Percent Range | >99% |
Molecular Formula | C23H36N6O5S |
Formula Weight | 508.63 |
Description
Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.This compound is a mixture of 2 diastereomers (approximate ratio 65/35).
Specifications
Argatroban | |
>99% | |
508.63 |
74863-84-6 | |
C23H36N6O5S |