missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argatroban, Tocris Bioscience™
Potent thrombin inhibitor
Brand: Tocris (Bio-Techne) 163750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity | 50mg |
---|
Description
Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.Chemical Identifiers
74863-84-6 | |
508.638 | |
argatroban, novastan, acova, argatroban inn:jan, argatrobanum, argatrobanum latin, argatroban anhydrous, mmtqap, mpqa, argatroban anhydrous | |
(2R,4R)-1-[(2S)-5-(diaminomethylideneamino)-2-[(3-methyl-1,2,3,4-tetrahydroquinolin-8-yl)sulfonylamino]pentanoyl]-4-methylpiperidine-2-carboxylic acid |
C23H36N6O5S | |
KXNPVXPOPUZYGB-IOVMHBDKSA-N | |
92722 | |
CC1CCN(C(C1)C(=O)O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C2=CC=CC3=C2NCC(C3)C |
Specifications
Argatroban | |
74863-84-6 | |
argatroban, novastan, acova, argatroban inn:jan, argatrobanum, argatrobanum latin, argatroban anhydrous, mmtqap, mpqa, argatroban anhydrous | |
CC1CCN(C(C1)C(=O)O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C2=CC=CC3=C2NCC(C3)C | |
508.638 | |
508.63 |
50mg | |
C23H36N6O5S | |
KXNPVXPOPUZYGB-IOVMHBDKSA-N | |
(2R,4R)-1-[(2S)-5-(diaminomethylideneamino)-2-[(3-methyl-1,2,3,4-tetrahydroquinolin-8-yl)sulfonylamino]pentanoyl]-4-methylpiperidine-2-carboxylic acid | |
92722 | |
>99% |