missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF10L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA555307
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ALEHSEEDGS IYEMADDPDV WVRSRPCARD AHRKEICSVA IISGGQGYRN FGSALGSSGR QAPCGETDST LLIWQVPL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
ARHGEF10L Polyclonal antibody specifically detects ARHGEF10L in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ARHGEF10L | |
Polyclonal | |
Unconjugated | |
ARHGEF10L | |
argal; Arhgef10l; GrinchGEF; KIAA1626; Rho guanine nucleotide exchange factor (GEF) 10-like; Rho guanine nucleotide exchange factor 10 like; rho guanine nucleotide exchange factor 10-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55160 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HCE6 | |
ARHGEF10L | |
Recombinant protein corresponding to Human ARGAL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only