missing translation for 'onlineSavingsMsg'
Learn More

Arecaidine propargyl ester tosylate, Tocris Bioscience™

Muscarinic agonist

Brand:  Tocris (Bio-Techne) 038310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13437603

  • £113.19 / 10mg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent muscarinic receptor agonist. Shows some selectivity for cardiac versus ileal M2 receptors.
Specifications

Chemical Identifiers

147202-94-6
351.417
arecaidine propargyl ester tosylate, tocris-0383, 4-methylbenzenesulfonic acid; prop-2-ynyl 1-methyl-3,6-dihydro-2h-pyridine-5-carboxylate, n-methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate, prop-2-yn-1-yl 1-methyl-5,6-dihydro-2h-pyridine-3-carboxylate; toluenesulfonic acid
4-methylbenzenesulfonic acid;prop-2-ynyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate
C17H21NO5S
LYHWIOMAWPVTPI-UHFFFAOYSA-N
6604936
CC1=CC=C(C=C1)S(=O)(=O)O.CN1CCC=C(C1)C(=O)OCC#C

Specifications

10mg
147202-94-6
arecaidine propargyl ester tosylate, tocris-0383, 4-methylbenzenesulfonic acid; prop-2-ynyl 1-methyl-3,6-dihydro-2h-pyridine-5-carboxylate, n-methyl-1,2,5,6-tetrahydropyridine-3-carboxylic acid propargyl ester tosylate, prop-2-yn-1-yl 1-methyl-5,6-dihydro-2h-pyridine-3-carboxylate; toluenesulfonic acid
CC1=CC=C(C=C1)S(=O)(=O)O.CN1CCC=C(C1)C(=O)OCC#C
351.417
351.42
Arecaidine propargyl ester tosylate
C17H21NO5S
LYHWIOMAWPVTPI-UHFFFAOYSA-N
4-methylbenzenesulfonic acid;prop-2-ynyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate
6604936
>98%
Videos
Documents