Learn More
Honeywell Safety Products™ Arctic Gloves
Handle frozen products safely and comfortably. Honeywell Safety Products™ Arctic Gloves can be used in cold storage conditions and by forklift operators and drivers.
Brand: Honeywell Safety Products™ 1640YCT8
Code : NEW
Additional Details : Weight : 0.14400kg
Description
- Grain goatskin.
- Water repellant Scotchguard™ treatment.
- Synthetic fur.
- Elasticized on back with zig-zag stitching.
- Good flexibility and outstanding dexterity.
- Supple.
- Good mechanical resistance.
Specifications
Dexterity: 5 | |
Elasticized back, Scotchguard™ treatment | |
30cm | |
Leather | |
1 | |
8 | |
Beige |
10cm | |
Agriculture, Building, Construction, Catering, Energy, Electricity, Food, Green Spaces, Iron, Steel, Logistics, Offshore Ship Building, Telecom, Utility and Wood industries | |
Yes | |
Synthetic fur | |
Cold | |
2 | |
CE CATEGORY: II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 511-2004 Cold hazards, EC Category PPE: 2, Quality Assurance: ISO 9001/2000, EC Certificate Number: N°0075/797/162/07/12/0587, EC Attestation Number: N°0075/797/162/07/12/0587 |