missing translation for 'onlineSavingsMsg'
Learn More

ARC 239 dihydrochloride, Tocris Bioscience™

α2B antagonist

Brand:  Tocris (Bio-Techne) 092810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13468163

  • £145.53 / 10mg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective α2B adrenoceptor antagonist (pKD values are 8.8, 6.7 and 6.4 at α2B, α2A, and α2D receptors respectively). Also available as part of the α2-Adrenoceptor Tocriset™ .
Specifications

Chemical Identifiers

55974-42-0
480.43
arc 239 dihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethyl-1,3-2h,4h-isoquinolindione dihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethylisoquinoline-1,3-dione dihydrochloride, arc239 dihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethyl-1,3-2h,4h-isoquinolindionedihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethylisoquinoline-1,3 2h,4h-dione dihydrochloride
2-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-4,4-dimethylisoquinoline-1,3-dione;dihydrochloride
C24H31Cl2N3O3
SAJKHRHHDGSJEZ-UHFFFAOYSA-N
10345075
CC1(C2=CC=CC=C2C(=O)N(C1=O)CCN3CCN(CC3)C4=CC=CC=C4OC)C.Cl.Cl

Specifications

10mg
55974-42-0
arc 239 dihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethyl-1,3-2h,4h-isoquinolindione dihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethylisoquinoline-1,3-dione dihydrochloride, arc239 dihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethyl-1,3-2h,4h-isoquinolindionedihydrochloride, 2-2-4-2-methoxyphenyl piperazin-1-yl ethyl-4,4-dimethylisoquinoline-1,3 2h,4h-dione dihydrochloride
CC1(C2=CC=CC=C2C(=O)N(C1=O)CCN3CCN(CC3)C4=CC=CC=C4OC)C.Cl.Cl
480.43
480.43
ARC 239 dihydrochloride
C24H31Cl2N3O3
SAJKHRHHDGSJEZ-UHFFFAOYSA-N
2-[2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl]-4,4-dimethylisoquinoline-1,3-dione;dihydrochloride
10345075
>98%
Videos
Documents