Learn More
ARAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557465
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: HCLEHKDDKL RNRPRKHRSF NCLEDTEPEA PLGQPKGHKG LKTLRKTEDR NSKATLDSDH KLPSRVIEEL NVVLQRSRTL PKELQDEQI Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
ARAP2 Polyclonal antibody specifically detects ARAP2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ARAP2 | |
Polyclonal | |
Unconjugated | |
ARAP2 | |
Arap2; Arf and Rho GAP adapter protein 2; arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 2; ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2; centaurin; centaurin, delta 1; centaurin-delta-1; CENTD1; Cnt-d1; Gm148; KIAA0580; mKIAA0580; PARX; PARX protein; Protein PARX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116984 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WZ64 | |
ARAP2 | |
Recombinant protein corresponding to Human ARAP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only