missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Aracut™ Grip Gloves

Catalog Number p-7115901
Click to view available options
Size:
6
7
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed with PVC patterned dots for improved abrasion resistance and grip. Honeywell Safety Products™ Aracut™ Grip Gloves are knitted for hand conformity and dexterity and constructed with Kevlar™ fiber for cutting resistance and thermal protection.

  • Knitted Kevlar™ glove.
  • Hand conformity and good dexterity.
  • Blue PVC dots on palm for grip and abrasion resistance.
  • Knitted construction for hand conformity and dexterity.
  • Excellent cutting resistance and protection against thermal hazards in a dry atmosphere.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-94 EEC Basic standards, EN 388-94 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/05/08/0134, EC Attestation Number 0075/797/162/05/08/0134
Coated Area Palm, Fingertips
Description Gauge 10
Coating Material PVC Dots
Color Yellow
Cut Resistance Rating 3
Material Aramid, PVC, Dupont™ Kevlar™ Aramid Fibers
Product Type Gloves
Size 11
For Use With (Application) Automotive, Building, Construction, Glass, Metal and Paper industries
Quantity 20/PK
Tear Rating 4
Length (Metric) 25 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'