missing translation for 'onlineSavingsMsg'
Learn More

AR-M 1000390 hydrochloride, Tocris Bioscience™

Low-internalizing δ agonist

Brand:  RD Systems (Bio-Techne) 4335/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13434574

  • £710.01 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Non-peptidic, low-internalizing δ-selective opioid receptor agonist; derivative of SNC 80 (Cat. No. 0764). Does not trigger acute desensitization of the analgesic response; reduces CFA-induced hyperalgesia. Brain penetrant following systemic administration.
Specifications

Chemical Identifiers

209808-47-9
384.948
ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride
N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride
C23H29ClN2O
OTXTZCLQEGSAMW-UHFFFAOYSA-N
76848958
CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl

Specifications

50mg
209808-47-9
ar-m 1000390 hydrochloride, arm390 hydrochloride, arm-390hcl, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl-benzamide hydrochloride, n,n-diethyl-4-phenyl piperidin-4-ylidene methyl benzamide hydrochloride, arm1000390 hydrochloride, n,n-diethyl-4-phenyl-4-piperidinylidenemethyl benzamide hydrochloride
CCN(CC)C(=O)C1=CC=C(C=C1)C(=C2CCNCC2)C3=CC=CC=C3.Cl
384.948
384.94
AR-M 1000390 hydrochloride
C23H29ClN2O
OTXTZCLQEGSAMW-UHFFFAOYSA-N
N,N-diethyl-4-[phenyl(piperidin-4-ylidene)methyl]benzamide;hydrochloride
76848958
>97%
Videos
Documents