missing translation for 'onlineSavingsMsg'
Learn More

Duran™ AR Glass Disposable Culture Tubes

Disposable culture tubes for general purpose. DURAN™ AR Glass Disposable Culture Tubes are used in the laboratory setting.

Brand:  Duran™ 231720587

Code : A0

Additional Details : Weight : 2.75000kg

 View more versions of this product

Product Code. 10208801

  • £33.20 / Pack of 550


Estimated Shipment
14-06-2018
Add to basket

Specifications

Specifications

550
5mL
AR Glass
550Pack
75mm
Disposable Culture Tube
Videos