Learn More
National Diagnostics™ AquaPor™ ES GTAC Agarose
Brand:  National Diagnostics™ EC-203-25
UNSPSC : 41105300
Code : LF
Additional Details : CAS Number : 9012-36-6 Weight : 0.01000kg
Product Code. 12311489
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
Premium, ultra-high strength, ultra-low EEO agarose, ideal for PFGE.
Specifications
Specifications
Agarose
Low
Free
Free
Pulsed field gel electrophoresis (PFGE)
25g
Documentation
Technical Documents