missing translation for 'onlineSavingsMsg'
Learn More

AQ-RA 741, Tocris Bioscience™

Selective, high affinity M2 antagonist

Brand:  Tocris (Bio-Techne) 229210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460954

  • £163.17 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

High affinity, selective muscarinic M2 receptor antagonist (pKi values are 8.3, 7.7 and 6.82 for M2, M1 and M3 receptors, respectively). Displays cardioselectivity in vivo, over intestinal, tracheal and bladder muscarinic receptors; inhibits vagally and agonist-induced bradycardia.
Specifications

Chemical Identifiers

123548-16-3
463.626
aq-ra 741, aq-ra-741, 11-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 6h-pyrido 2,3-b 1,4 benzodiazepin-6-one,11-2-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro, acmc-20ej93, d0vz3d, aq-ra-741bs, chembl43383, 11-2-4-4-diethylamino butyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-one
11-[2-[4-[4-(diethylamino)butyl]piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
C27H37N5O2
BCUGCHZRMKTPMU-UHFFFAOYSA-N
129989
CCN(CC)CCCCC1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4

Specifications

10mg
123548-16-3
aq-ra 741, aq-ra-741, 11-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro-6h-pyrido 2,3-b 1,4 benzodiazepin-6-one, 6h-pyrido 2,3-b 1,4 benzodiazepin-6-one,11-2-4-4-diethylamino butyl-1-piperidinyl acetyl-5,11-dihydro, acmc-20ej93, d0vz3d, aq-ra-741bs, chembl43383, 11-2-4-4-diethylamino butyl piperidin-1-yl acetyl-5h-benzo e pyrido 3,2-b 1,4 diazepin-6 11h-one
CCN(CC)CCCCC1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4
463.626
463.62
AQ-RA 741
C27H37N5O2
BCUGCHZRMKTPMU-UHFFFAOYSA-N
11-[2-[4-[4-(diethylamino)butyl]piperidin-1-yl]acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one
129989
>99%
Videos
Documents