missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Drosophila Vials

Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging

Brand:  Fisherbrand™

UNSPSC : 41102501

Code : 33

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12766075

  • £93.89 / Pack of 500


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

Yes
28.5mm
95mm
Wide
Clear
Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Polypropylene
Fly Drosophila Vial
Documentation View accessory products