Learn More
Fisherbrand™ Drosophila Vials
Straight wall design minimizes the dislodging of food; provides more efficient racking, filling and plugging
Brand:  Fisherbrand™
Code : 33
Additional Details : Weight : 5.32970kg
 View more versions of this product
Product Code. 12736075
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
Fisherbrand Drosophila vials are available in clear polystyrene, translucent and flexible polypropylene, and virtually unbreakable K-resin. These vials feature a flat bottom, straight wall design for convenient racking, filling and stacking. Available bulk packaged or in a convenient, 10 × 10 tray pack configuration.
Polypropylene Vials
  • Translucent and flexible
  • Autoclavable
Polystyrene Vials
  • Excellent clarity for optimal visibility
  • Nonautoclavable
K-Resin Vials
  • Highly flexible, virtually unbreakable
  • Nonautoclavable
Standard-Packed Glass Shell Vials
  • Type 1, N-51A borosilicate glass
  • Securely packed in cardboard trays with individual vial dividers
Bulk- and Tray-Packed Shell Vials
  • Available in narrow- or wide-mouth styles
  • Constructed from polystyrene, K-Resin™, or polypropylene
  • K-Resin and polypropylene vials are virtually unbreakable, scratch and mar resistant; polystyrene and K-Resin vials have glass-like clarity
  • Polypropylene vials are autoclavable; polystyrene and K-Resin are nonautoclavable
Specifications
Specifications
No
28.5mm
95mm
Narrow
Clear
Straight wall design minimizes the dislodging of food, provides more efficient racking, filling and plugging
Polystyrene
Drosophila Vial, Tray Packed
Videos
Documentation View accessory products