Learn More
Fisherbrand™ Drosophila Products, BuzzPlugs™ Vial and Bottle Closures
Nonshedding, dense weave of nontoxic cellulose acetate minimizes evaporation, keeps food fresh longer
Brand:  Fisherbrand™
Code : 33
Additional Details : Weight : 1.92000kg
 View more versions of this product
Product Code. 12796075
  • £125.40 / Pack of 1000
In Stock 40
Add to basket
You may only order up to the available stock. For more details call customer services on 01509 555 500
Specifications
Specifications
Minimizes evaporation, keeps food fresh longer
Cellulose Acetate
Standard (2.4cm dia.) glass vials
Superior barrier
1000 Case
BuzzPlugs™ Bottle Closure
Videos
Documentation