Learn More
TaqMan™, GTXpress™ TaqMan™ GTXpress™ Master Mix
Combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format.
Brand: Applied Biosystems™ 4401857
Code : 50
Additional Details : Weight : 0.00450kg
Description
TaqMan™ GTXpress™ Master Mix delivers accurate genotyping results with robust performance in less than 50 min. The 100mL pack (2 x 50mL) includes reagents sufficient to perform 40,000 PCR reactions (5μL/reaction).
The TaqMan™ GTXpress™ Master Mix delivers accurate genotyping results with robust performance in less than 50 min. Applied Biosystems™ TaqMan™ GTXpress™ Master Mix (2X) combines AmpliTaq™ Fast DNA Polymerase, UP, dNTPs, Tracking Dye, and ROX™ dye in a convenient single vial format. The 100 mL pack (2 x 50 mL) includes reagents sufficient to perform 40,000 PCR reactions (5μL/reaction).Specifications
High | |
2 x 50 mL | |
2X |
qPCR | |
Genotyping |
For Research Use Only. Not for use in diagnostic procedures.