missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ MicroAmp™ EnduraPlate™ Optical 96-Well Clear GPLE Reaction Plates with Barcode
Each lot is certified DNA, RNAse, and PCR inhibitor-free
Brand: Applied Biosystems™ 4483351
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- For Laboratory Use
- Optimized for performance with Applied Biosystems instruments
- High mechanical stability and flatness for complete workflow needs
- Polypropylene and high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate offers mechanical stability, flatness, and high durability
- ANSI™/SBS-compliant construction
- Ideal choice for use with automation/robotics and multi-instrument experiments (like thermal cycler, real-time thermal cycler, genetic analyzer)
- Thin-wall construction allows for snug fit to thermal blocks with high heat transfer from thermal block to sample
- Easy-to-read, black, well-identification text helps to minimize pipetting mistakes and improves sample preparation productivity
- Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System
Fast PCR, PCR, PCR & Real-Time PCR, Routine PCR
Specifications
Includes Barcode | |
500 96-Well plates. Store at room temperature. | |
MicroAmp | |
Microplate | |
Semi | |
0.3 mL | |
0.2 mL | |
Fast, Standard |
Certified DNA/RNase Free | |
500 Plates | |
Optical, Clear | |
Thermal Cycler, Genetic Analyzer, DNA Analyzer | |
Polycarbonate, Polypropylene | |
96 | |
500 Plates |
For Laboratory Use