Learn More
Applied Biosystems™ MicroAmp™ 96-Well Full Plate Cover
Seal up to 96-wells in MicroAmp Tray/Retainer Set or Reaction Plates to prevent evaporation during thermal cycling of samples
Brand:  Applied Biosystems™ N8010550
Code : 50
Additional Details : Weight : 0.13000kg
Product Code. 10732897
  • £111.00 / Each
Estimated Shipment: 02-05-2024
to see stock.
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
  • Durable design allows for continuous use with high-quality results
  • Convenient, cost-saving alternative to MicroAmp Caps
  • Patented one-piece design for convenience
  • Reusable up to five times
  • Manufactured from PCR-compatible rubber
  • Non-sterile and autoclavable
  • Platform: GeneAmp™ 9700
See user's manual or package insert for limited label license, and trademark information.
PCR, PCR & Real-Time PCR
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
Certified DNA/RNase Free
5 Covers
Natural
2720 Thermal Cycler, 6100 Thermal Cycler, GeneAmp 9700, 2720 Thermal Cycler
Non-sterile
96-Well Plates
MicroAmp™ 96-Well Full Plate Cover is manufactured from PCR-compatible rubber. It is non-sterile, autoclavable and guaranteed reusable up to five times.
MicroAmp
Microplate Lid
96
PCR
Room Temperature