Learn More
Applied Biosystems™ MagMAX™ Express Tip Combs
For use with MagMAX Express Magnetic Particle Processors
Brand: Applied Biosystems™ 4388452
Code : 50
Additional Details : Weight : 4.05000kg
Includes: 50 combs
Description
- Fit models 4400074 and 4400075
Animal Health, Animal Health Sample Preparation, Automated Nucleic Acid Purification, Automated Total RNA Isolation, Automated Viral DNA/RNA Purification, DNA & RNA Purification & Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells & Tissues, Total RNA from Bacterial Samples, Total RNA from Blood, Viral RNA/DNA Purification
Order Info
Shipping Condition: Room Temperature
Specifications
Filtered | |
Store at room temperature. | |
MagMAX™ Express Magnetic Particle Processor |
Universal Fit | |
Room Temperature | |
MagMAX |
For Research Use Only. Not for use in diagnostic procedures.