missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AmpliTaq Gold™ 360 DNA Polymerase

Efficiently and reproducibly amplifies long (up to 5 Kb) sequences

Brand:  Applied Biosystems™ 4398898

15798.50 GBP valid until 2024-12-31
Use promo code "21967" to get your promotional price.



Code : 50

Additional Details : CAS Number : 7732-18-5 Weight : 0.50000kg

 View more versions of this product

Product Code. 10525405

  • £17340.00 / 25000 units
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

pplied Biosystems™ AmpliTaq Gold™ 360 DNA Polymerase, when used with improved AmpliTaq Gold 360 Buffer, and optional 360 GC Enhancer, amplifies vast range of DNA sequence contexts. AmpliTaq Gold™ 360 DNA Polymerase delivers 360° coverage for a full range of targets

  • Optimized for broadest range of targets–from everyday to challenging
  • Unmatched sensitivity, specificity, and yield
  • Robust amplification of GC-rich sequences with market-leading 360 GC Enhancer
  • Achieves highest quality sequencing data
  • Less waste, sustainable packaging

See user's manual or package insert for limited label license, and trademark information.

Amplification of Bisulfite-Treated DNA, GC-Rich PCR, Hot Start PCR, Methylation Analysis, PCR, PCR and Real-Time PCR, RNAi, Epigenetics and Non-Coding RNA Research, Routine PCR

Specifications

Specifications

• 25 x 1000 U AmpliTaq Gold 360 DNA Polymerase
• 100 x 1.5 mL AmpliTaq Gold 360 Buffer, 10X
• 100 x 1.5 mL 25 mM MgCl2
• 100 x 1.5 mL 360 GC Enhancer

Store at -15 to -25°C.
Tube
AmpliTaq Gold 360 DNA Polymerase
5' - 3'
AmpliTaq Gold™
25 x 1000 Units
1 X
20000
Separate Components
DNA
Primer-probe
High
Standard
Sustainable packaging
DNA Polymerase
5 U/μL
Built-In Hot Start
3'-A
5 kb or less
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.