missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Apoptosis Inhibitor II, Ns3694

Inhibits apoptosome formation

Brand:  Alfa Aesar™ J65987.MA

Code : NEW

Additional Details : CAS Number : 426834-38-0 Weight : 0.00010kg

 View more versions of this product

Product Code. 15487169

  • £92.22 / 10mg


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Specifications

10mg
2° to 8°C
Inhibits apoptosome formation
C15H10ClF3N2O3
4-Chloro-2-(3-[3-(trifluoromethyl)phenyl]ureido)benzoic acid
White
Powder
426834-38-0
358.70g/mol
Signal Transduction Reagent
Videos
SDS