missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOBEC3D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563066
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: VFRGPVLPKR QSNHRQEVYF RFENHAEMCF Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 33%.
APOBEC3D Polyclonal antibody specifically detects APOBEC3D in Human samples. It is validated for Immunohistochemistry.Specifications
APOBEC3D | |
Polyclonal | |
Unconjugated | |
APOBEC3D | |
A3D; APOBEC3D; APOBEC3DE; APOBEC3E; apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3D; apolipoprotein B mRNA editing enzyme catalytic subunit 3D; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3D; apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3E pseudogene; ARP6; DNA dC->dU-editing enzyme APOBEC-3D; probable DNA dC->dU-editing enzyme APOBEC-3D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140564 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AK3 | |
APOBEC3D | |
Recombinant protein corresponding to Human APOBEC3D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only