missing translation for 'onlineSavingsMsg'
Learn More

Apigenin, Tocris Bioscience™

Protein kinase inhibitor

Brand:  Tocris (Bio-Techne) 122750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13428653

  • £166.11 / 50mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Protein kinase inhibitor. Suppresses tumor-promoting effects of TPA and exhibits antiproliferative activity in human breast cancer cells (IC50 values are 59.44 and 31.15 μM at 24 and 72 hrs respectively). Activates both the intrinsic and extrinsic apoptotic pathways and displays anti-inflammatory, hypotensive, antispasmodic and antioxidant properties in vivo.
Specifications

Chemical Identifiers

520-36-5
270.24
apigenin, 5,7-dihydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, chamomile, spigenin, versulin, apigenol, 4',5,7-trihydroxyflavone, apigenine, c.i. natural yellow 1, 5,7,4'-trihydroxyflavone
CHEBI:18388
C1=CC(=CC=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O
C15H10O5
KZNIFHPLKGYRTM-UHFFFAOYSA-N
5280443
5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one

Specifications

50mg
520-36-5
apigenin, 5,7-dihydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, chamomile, spigenin, versulin, apigenol, 4',5,7-trihydroxyflavone, apigenine, c.i. natural yellow 1, 5,7,4'-trihydroxyflavone
C1=CC(=CC=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O
270.24
CHEBI:18388
>98%
Apigenin
C15H10O5
KZNIFHPLKGYRTM-UHFFFAOYSA-N
5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
5280443
270.2
Videos
Documents