Learn More
Fisherbrand™ Apiezon Grease
Lubricate pumps with this grease. Fisherbrand™ Apiezon Grease is designed for use with Fisherbrand™ rotary vane pumps.
Brand:  Fisherbrand™
Code : 03
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12981141
  • £235.67 / 25g
Estimated Shipment
19-06-2018
Add to basket
Description
Description
  • Apiezon grease
  • For general use in Fisherbrand rotary vane pumps
Specifications
Specifications
Apiezon™ Grease
25g
Videos
Documentation