missing translation for 'onlineSavingsMsg'
Learn More

Apatite, naturally occurring mineral, grains, approximately 0.06-0.19in, Thermo Scientific Chemicals

Catalog Number 11319128
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1306-05-4
Ca5FO12P3
504.298
MFCD00285511
VSIIXMUUUJUKCM-UHFFFAOYSA-D
fluorapatite, apatite, phosphate rock, unii-m4cm1h238j, phospate rock, fluorapatite ca5f po4 3
10207414
pentacalcium;fluoride;triphosphate
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[F-].[Ca+2].[Ca+2].[Ca+2].[Ca+2].[Ca+2]
This item is not returnable. View return policy
1306-05-4
Ca5FO12P3
504.298
MFCD00285511
VSIIXMUUUJUKCM-UHFFFAOYSA-D
fluorapatite, apatite, phosphate rock, unii-m4cm1h238j, phospate rock, fluorapatite ca5f po4 3
10207414
pentacalcium;fluoride;triphosphate
[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[F-].[Ca+2].[Ca+2].[Ca+2].[Ca+2].[Ca+2]

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Chips
Quantity 1 g
Solubility Information Soluble in water and hydrochloric acid.
Appearance Colorless chips
Chemical Name or Material Apatite

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'