missing translation for 'onlineSavingsMsg'
Learn More

AP 24534, Tocris Bioscience™

Potent multi-kinase and pan-Bcr-Abl inhibitor

Brand:  Tocris (Bio-Techne) 427450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444474

  • £998.13 / 50mg

Estimated Shipment date
from Supplier 28-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent multi-kinase and pan-Bcr-Abl inhibitor. Displays potent activity against cell lines expressing native Bcr-Abl or Bcr-AblT3151 (IC50 values are 0.37 and 2.0 nM respectively); also inhibits other Abl kinase domain mutants at nanomolar potencies. Exhibits inhibitory activity against PDGFRα, c-Src and c-Kit (IC50 values are 1.1, 5.4 and 12.5 nM respectively); potently inhibits FGFR and VEGFR family kinases. Orally active.
Specifications

Chemical Identifiers

943319-70-8
532.571
ponatinib, ponatinib ap24534, iclusig, 3-imidazo 1,2-b pyridazin-3-ylethynyl-4-methyl-n-4-4-methylpiperazin-1-yl methyl-3-trifluoromethyl phenyl benzamide, ap24534 ponatinib, iclusig tn, 3-2-imidazo 1,2-b pyridazin-3-yl ethynyl-4-methyl-n-4-4-methylpiperazin-1-yl methyl-3-trifluoromethyl phenyl benzamide, benzamide, 3-2-imidazo 1,2-b pyridazin-3-ylethynyl-4-methyl-n-4-4-methyl-1-piperazinyl methyl-3-trifluoromethyl phenyl, ponatinib usan:inn
CHEBI:78543
CC1=C(C=C(C=C1)C(=O)NC2=CC(=C(C=C2)CN3CCN(CC3)C)C(F)(F)F)C#CC4=CN=C5N4N=CC=C5
C29H27F3N6O
PHXJVRSECIGDHY-UHFFFAOYSA-N
24826799
3-(2-imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-[4-[(4-methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)phenyl]benzamide

Specifications

50mg
943319-70-8
ponatinib, ponatinib ap24534, iclusig, 3-imidazo 1,2-b pyridazin-3-ylethynyl-4-methyl-n-4-4-methylpiperazin-1-yl methyl-3-trifluoromethyl phenyl benzamide, ap24534 ponatinib, iclusig tn, 3-2-imidazo 1,2-b pyridazin-3-yl ethynyl-4-methyl-n-4-4-methylpiperazin-1-yl methyl-3-trifluoromethyl phenyl benzamide, benzamide, 3-2-imidazo 1,2-b pyridazin-3-ylethynyl-4-methyl-n-4-4-methyl-1-piperazinyl methyl-3-trifluoromethyl phenyl, ponatinib usan:inn
CC1=C(C=C(C=C1)C(=O)NC2=CC(=C(C=C2)CN3CCN(CC3)C)C(F)(F)F)C#CC4=CN=C5N4N=CC=C5
532.571
CHEBI:78543
>98%
AP 24534
C29H27F3N6O
PHXJVRSECIGDHY-UHFFFAOYSA-N
3-(2-imidazo[1,2-b]pyridazin-3-ylethynyl)-4-methyl-N-[4-[(4-methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)phenyl]benzamide
24826799
532.56
Videos
Documents