missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ Alkaline Phosphatase

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Brand:  Invitrogen™ IVGN2204

Code : NEW

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 15545720

  • £79.94 / 1 set


EU Stock 6
Add to basket

Includes: Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Description

Description

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• Utilizes the Anza 10X Buffer
• One-step digestion and dephosphorylation protocol
• Two-step dephosphorylated protocols available
• Heat inactivated in 5 minutes

Certifications

CoA

Specifications

Specifications

1U/μL
500 reactions
Store at -20°C
Anza™ Alkaline Phosphatase, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Cloning, Gene Expression/RNA Analysis, Protein Expression, Genotyping and Genetic Variation, RNAi, Sequencing
Videos

For research use only. Not for use in diagnostic procedures.