missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Anza™ 25 PaeI

Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.

Brand:  Invitrogen™ IVGN0254

Code : NEW

Additional Details : Weight : 0.40000kg

 View more versions of this product

Product Code. 15585710

  • £56.50 / 1 set

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer

Description

Description

The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.
• One buffer for all restriction enzymes
• One digestion protocol for all DNA types
• Complete digestion in 15 min
• Overnight digestion without star activity

Certifications

CoA

Specifications

Specifications

400U
Approved for shipment on Wet or Dry Ice
Not dam methylation-sensitive, Not dcm methylation-sensitive, Not CpG methylation-sensitive
Anza™ 10x Red Buffer, Anza™ 10x Buffer
37°C
40 µL Anza 25 PaeI
500 µL Anza Buffer (10X)
500 µL Anza Red Buffer (10X)

Store at -20°C.
Anza™
PaeI
Yes
Videos
Documents

For research use only. Not for use in diagnostic procedures.