missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Antireproductive tripeptide

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J64717.MB

Code : NEW

Additional Details : CAS Number : 71730-64-8 Weight : 0.00025kg

 View more versions of this product

Product Code. 15436539

  • £222.92 / 25mg


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Specifications

334.37
25mg
H-Thr-Ser-Lys-OH
-30° to -10°C
Antireproductive tripeptide
HPLC
Powder
RUO
Bovine Pineal Antireproductive Tripeptide
Videos
SDS

For Research Use Only