missing translation for 'onlineSavingsMsg'
Learn More

Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048298
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]
This item is not returnable. View return policy
7440-36-0
Sb
121.76
MFCD00011211 MFCD00134030
WATWJIUSRGPENY-UHFFFAOYSA-N
powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes german
5354495
CHEBI:30304
antimony
[Sb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1380°C
UN Number UN2871
Quantity 250 g
Percent Purity 99.5%
Mesh Size −100 Mesh
Odor Odorless
Weight Apparent density: 2.4g/cm3 to 2.9g/cm3
Form Powder
Assay (metals basis)
Chemical Name or Material Antimony powder
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Antimony powder
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Acute toxicity Category 4
Hazard Statement
  • H301-Toxic if swallowed.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'