missing translation for 'onlineSavingsMsg'
Learn More

Antimony potassium tartrate hydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7081069
Click to view available options
Quantity:
100 g
500 g
2500 g
331753-56-1
C8H4K2O12Sb2
613.83
MFCD00148863
GUJUCWZGYWASLH-UHFFFAOYNA-J
potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
331753-56-1
C8H4K2O12Sb2
613.83
MFCD00148863
GUJUCWZGYWASLH-UHFFFAOYNA-J
potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 2.607 g/mL
Assay Percent Range 98%
Quantity 100 g
UN Number UN1551
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 613.82 (anhydrous)
Percent Purity 98%
Chemical Name or Material Antimony potassium tartrate hydrate
compliance-icons compliance-icons
Product Identifier
  • Potassium antimonyl tartrate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'